Assay Design
Gene Transcripts
- Gene Symbol
- csnk1db
- Entrez Gene ID
-
406533
- Gene Name
- casein kinase 1, delta b
- Gene Aliases
-
csnk1d, wu:fb69b03, wu:fj97a05, zgc:77627
- Chromosome Location
-
Chr.12: 33320537 - 33336787 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_213250.1 |
NP_998415.1 |
7 - 8 |
1424 |
60 |
GenBank mRNA |
BC063953.1 |
- |
7 - 8 |
1424 |
60 |
View Full Product Details