Assay Design



Gene Transcripts

Gene Symbol
rps13
Entrez Gene ID
415169
Gene Name
ribosomal protein S13
Gene Aliases
fb14f11, fd13f08, fd60g01, wu:fb14f11, wu:fd13f08, wu:fd60g01, zgc:91809
Chromosome Location
Chr.7: 72671 - 75308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002079.1 NP_001002079.1 3 - 4 158 68
GenBank mRNA BC072552.1 - 3 - 4 158 68
BC163978.1 - 3 - 4 182 68

View Full Product Details