Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002211.1 | NP_001002211.1 | 7 - 8 | 886 | 66 |
XM_005155296.4 | XP_005155353.1 | 7 - 8 | 947 | 66 | |
XM_005155298.3 | XP_005155355.1 | 7 - 8 | 883 | 66 | |
GenBank mRNA | BC074073.1 | - | 7 - 8 | 886 | 66 |
BC164605.1 | - | 6 - 7 | 668 | 66 |