Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012385.1 | NP_001012385.1 | 1 - 2 | 220 | 99 |
XM_005156825.4 | XP_005156882.1 | 1 - 2 | 266 | 99 | |
GenBank mRNA | AJ698458.1 | - | 1 - 2 | 220 | 99 |
AJ717297.1 | - | 1 - 2 | 220 | 99 |