Assay Design



Gene Transcripts

Gene Symbol
matn3b
Entrez Gene ID
497645
Gene Name
matrilin 3b
Gene Aliases
zgc:123028
Chromosome Location
Chr.13: 32627283 - 32635905 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012385.1 NP_001012385.1 3 - 4 1200 87
XM_005156826.4 XP_005156883.1 2 - 3 831 87
GenBank mRNA AJ698730.2 - 2 - 3 816 87
AJ717297.1 - 3 - 4 1200 87

View Full Product Details