Assay Design



Gene Transcripts

Gene Symbol
snrpd2
Entrez Gene ID
550244
Gene Name
small nuclear ribonucleoprotein D2 polypeptide
Gene Aliases
im:6908977, si:dkey-113g17.2, zgc:110732
Chromosome Location
Chr.18: 98443 - 106640 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017582.1 NP_001017582.1 2 - 3 269 104
GenBank mRNA BC093003.1 - 2 - 3 269 104

View Full Product Details