Assay Design



Gene Transcripts

Gene Symbol
cfap52
Entrez Gene ID
553666
Gene Name
cilia and flagella associated protein 52
Gene Aliases
fj15f10, si:dkey-28k21.1, wdr16, wu:fj15f10, zgc:111988
Chromosome Location
Chr.6: 312137 - 328005 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020639.1 NP_001018475.1 12 - 13 1607 93
GenBank mRNA BC095635.1 - 12 - 13 1607 93

View Full Product Details