Assay Design



Gene Transcripts

Gene Symbol
emilin1a
Entrez Gene ID
566473
Gene Name
elastin microfibril interfacer 1a
Gene Aliases
im:7159005, si:dkey-20n10.2
Chromosome Location
Chr.20: 34845657 - 34865018 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030207.1 NP_001025378.1 4 - 5 2439 107
XM_005160756.3 XP_005160813.1 4 - 5 2779 107

View Full Product Details