Assay Design



Gene Transcripts

Gene Symbol
ppil1
Entrez Gene ID
558042
Gene Name
peptidylprolyl isomerase (cyclophilin)-like 1
Gene Aliases
zgc:123245
Chromosome Location
Chr.6: 40647788 - 40651935 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034178.1 NP_001029350.1 1 - 2 177 70
GenBank mRNA BC105964.1 - 1 - 2 177 70

View Full Product Details