Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034975.1 | NP_001030147.1 | 28 - 29 | 3039 | 69 |
XM_009296502.3 | XP_009294777.1 | 28 - 29 | 3160 | 69 | |
XM_021469336.1 | XP_021325011.1 | 28 - 29 | 3166 | 69 | |
GenBank mRNA | BC163568.1 | - | 28 - 29 | 3027 | 69 |
CU638802.1 | - | 28 - 29 | 3052 | 69 | |
DQ153065.1 | - | 28 - 29 | 3039 | 69 |