Assay Design



Gene Transcripts

Gene Symbol
srek1ip1
Entrez Gene ID
553315
Gene Name
SREK1-interacting protein 1
Gene Aliases
sfrs12ip1, zgc:123019
Chromosome Location
Chr.10: 11394528 - 11397559 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037672.1 NP_001032761.1 2 - 3 135 92
GenBank mRNA BC107616.1 - 2 - 3 135 92

View Full Product Details