Assay Design



Gene Transcripts

Gene Symbol
slc25a47a
Entrez Gene ID
724009
Gene Name
solute carrier family 25 member 47a
Gene Aliases
hdmcpa, slc25a47, zgc:136752
Chromosome Location
Chr.20: 53627848 - 53641881 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045314.1 NP_001038779.1 4 - 5 361 106
XM_017352527.2 XP_017208016.2 5 - 6 946 106
XM_021468518.1 XP_021324193.1 5 - 6 968 106
XM_021468519.1 XP_021324194.1 5 - 6 946 106
GenBank mRNA BC117643.1 - 4 - 5 361 106

View Full Product Details