Assay Design



Gene Transcripts

Gene Symbol
slc25a47a
Entrez Gene ID
724009
Gene Name
solute carrier family 25 member 47a
Gene Aliases
hdmcpa, slc25a47, zgc:136752
Chromosome Location
Chr.20: 53627848 - 53641881 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045314.1 NP_001038779.1 5 - 6 634 89
XM_021468518.1 XP_021324193.1 6 - 7 1241 89
GenBank mRNA BC117643.1 - 5 - 6 634 89

View Full Product Details