Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079834.1 | NP_001073303.1 | 2 - 3 | 325 | 64 |
GenBank mRNA | BC150218.1 | - | 2 - 3 | 604 | 64 |
BC163085.1 | - | 2 - 3 | 320 | 64 | |
BC163092.1 | - | 2 - 3 | 320 | 64 | |
DQ465921.1 | - | 2 - 3 | 325 | 64 |