Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098392.1 | NP_001091862.1 | 2 - 3 | 887 | 67 |
GenBank mRNA | BC162315.1 | - | 2 - 3 | 821 | 67 |
BC162318.1 | - | 2 - 3 | 821 | 67 | |
EF550998.1 | - | 2 - 3 | 887 | 67 | |
JN615142.1 | - | 2 - 3 | 852 | 67 | |
JN809237.1 | - | 2 - 3 | 714 | 67 |