Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_131057.1 | NP_571132.1 | 11 - 12 | 514 | 94 |
XM_005156453.4 | XP_005156510.1 | 11 - 12 | 529 | 94 | |
XM_017357926.2 | XP_017213415.1 | 11 - 12 | 529 | 94 | |
XM_017357927.2 | XP_017213416.1 | 11 - 12 | 529 | 94 | |
XM_021479102.1 | XP_021334777.1 | 10 - 11 | 490 | 94 | |
XM_021479103.1 | XP_021334778.1 | 9 - 10 | 396 | 94 | |
XM_021479104.1 | XP_021334779.1 | 10 - 11 | 473 | 94 | |
XM_021479105.1 | XP_021334780.1 | 9 - 10 | 396 | 94 | |
GenBank mRNA | AB005147.1 | - | 11 - 12 | 514 | 94 |
BC129275.1 | - | 10 - 11 | 481 | 94 | |
Y12007.1 | - | - | 431 | 94 |