Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_131057.1 | NP_571132.1 | 24 - 25 | 1757 | 125 |
XM_005156453.4 | XP_005156510.1 | 24 - 25 | 1769 | 125 | |
XM_017357926.2 | XP_017213415.1 | 24 - 25 | 1766 | 125 | |
XM_017357927.2 | XP_017213416.1 | 23 - 24 | 1736 | 125 | |
XM_021479102.1 | XP_021334777.1 | 23 - 24 | 1733 | 125 | |
XM_021479103.1 | XP_021334778.1 | 22 - 23 | 1636 | 125 | |
XM_021479104.1 | XP_021334779.1 | 22 - 23 | 1680 | 125 | |
XM_021479105.1 | XP_021334780.1 | 21 - 22 | 1600 | 125 | |
GenBank mRNA | AB005147.1 | - | 24 - 25 | 1757 | 125 |
BC129275.1 | - | 22 - 23 | 1688 | 125 |