Assay Design



Gene Transcripts

Gene Symbol
mmp14a
Entrez Gene ID
373882
Gene Name
matrix metallopeptidase 14a (membrane-inserted)
Gene Aliases
cb386, sb:cb386, wu:fb72b09
Chromosome Location
Chr.7: 24124298 - 24149631 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194416.1 NP_919397.1 5 - 6 1154 71
GenBank mRNA AY324196.1 - 5 - 6 1154 71
BC131864.1 - 5 - 6 1146 71

View Full Product Details