Assay Design



Gene Transcripts

Gene Symbol
fbp1a
Entrez Gene ID
335231
Gene Name
fructose-1,6-bisphosphatase 1a
Gene Aliases
fbp1, fbp1l, fk92h02, wu:fk92h02, zgc:64096
Chromosome Location
Chr.5: 44833313 - 44843738 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199942.2 NP_956236.1 1 - 2 337 106
GenBank mRNA BC057430.1 - - 247 106
EH585004.1 - 1 - 2 295 106

View Full Product Details