Assay Design
Gene Transcripts
- Gene Symbol
- fbp1a
- Entrez Gene ID
-
335231
- Gene Name
- fructose-1,6-bisphosphatase 1a
- Gene Aliases
-
fbp1, fbp1l, fk92h02, wu:fk92h02, zgc:64096
- Chromosome Location
-
Chr.5: 44833313 - 44843738 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_199942.2 |
NP_956236.1 |
5 - 6 |
865 |
72 |
GenBank mRNA |
BC057430.1 |
- |
- |
775 |
72 |
View Full Product Details