Assay Design
Gene Transcripts
- Gene Symbol
- tpst1l
- Entrez Gene ID
-
393171
- Gene Name
- tyrosylprotein sulfotransferase 1, like
- Gene Aliases
-
zgc:56162
- Chromosome Location
-
Chr.21: 31232873 - 31241164 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200202.2 |
NP_956496.1 |
2 - 3 |
1303 |
72 |
GenBank mRNA |
AF510737.1 |
- |
2 - 3 |
1303 |
72 |
View Full Product Details