Assay Design



Gene Transcripts

Gene Symbol
vil1
Entrez Gene ID
393207
Gene Name
villin 1
Gene Aliases
vil1l, zgc:55779
Chromosome Location
Chr.9: 44910768 - 44953424 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200238.1 NP_956532.1 11 - 12 1289 70
XM_005167920.4 XP_005167977.1 11 - 12 1367 70
GenBank mRNA BC047186.1 - 11 - 12 1289 70
BC165692.1 - 10 - 11 1242 70

View Full Product Details