Assay Design
Gene Transcripts
- Gene Symbol
- pcolce2b
- Entrez Gene ID
-
393432
- Gene Name
- procollagen C-endopeptidase enhancer 2b
- Gene Aliases
-
zgc:63590
- Chromosome Location
-
Chr.24: 5871589 - 5889763 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200460.1 |
NP_956754.1 |
2 - 3 |
250 |
82 |
GenBank mRNA |
BC055153.1 |
- |
2 - 3 |
250 |
82 |
View Full Product Details