Assay Design
Gene Transcripts
- Gene Symbol
- c8g
- Entrez Gene ID
-
393837
- Gene Name
- complement component 8, gamma polypeptide
- Gene Aliases
-
zgc:77874
- Chromosome Location
-
Chr.24: 17031129 - 17039617 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_200863.1 |
NP_957157.1 |
5 - 6 |
659 |
110 |
GenBank mRNA |
BC062865.1 |
- |
5 - 6 |
659 |
110 |
View Full Product Details