Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_200953.2 | NP_957247.1 | 14 - 15 | 1561 | 110 |
XM_009307227.3 | XP_009305502.1 | 14 - 15 | 1998 | 110 | |
GenBank mRNA | BC055591.1 | - | 14 - 15 | 1562 | 110 |
BC165713.1 | - | 13 - 14 | 1521 | 110 |