Assay Design



Gene Transcripts

Gene Symbol
eif4a3
Entrez Gene ID
394053
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
ddx48, eIF4A-III, zgc:56139
Chromosome Location
Chr.11: 44880191 - 44897708 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201078.1 NP_957372.1 5 - 6 604 104
GenBank mRNA BC045939.1 - 5 - 6 604 104
BC165833.1 - 5 - 6 524 104

View Full Product Details