Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201161.1 | NP_957455.1 | 11 - 12 | 948 | 87 |
XM_021468025.1 | XP_021323700.1 | 12 - 13 | 1074 | 87 | |
XM_021468026.1 | XP_021323701.1 | 11 - 12 | 1079 | 87 | |
GenBank mRNA | BC049418.1 | - | 11 - 12 | 948 | 87 |
BC097041.1 | - | 11 - 12 | 1064 | 87 |