Assay Design
Gene Transcripts
- Gene Symbol
- parp16
- Entrez Gene ID
-
405842
- Gene Name
- poly (ADP-ribose) polymerase family, member 16
- Gene Aliases
-
zgc:77744
- Chromosome Location
-
Chr.25: 31868316 - 31882618 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_212906.1 |
NP_998071.1 |
6 - 7 |
897 |
138 |
GenBank mRNA |
BC067335.1 |
- |
6 - 7 |
897 |
138 |
View Full Product Details