Assay Design
Gene Transcripts
- Gene Symbol
- arih1l
- Entrez Gene ID
-
405859
- Gene Name
- ariadne homolog, ubiquitin-conjugating enzyme E2 binding protein, 1 like
- Gene Aliases
-
wu:fc12f04, zgc:85905
- Chromosome Location
-
Chr.7: 155369 - 175687 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_212923.1 |
NP_998088.1 |
13 - 14 |
1574 |
138 |
GenBank mRNA |
BC067684.1 |
- |
13 - 14 |
1574 |
138 |
View Full Product Details