Assay Design



Gene Transcripts

Gene Symbol
cd9b
Entrez Gene ID
406737
Gene Name
CD9 molecule b
Gene Aliases
cd9l, fb82b11, wu:fb82b11, zgc:56512
Chromosome Location
Chr.4: 4234994 - 4261623 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213428.2 NP_998593.1 1 - 2 186 92
GenBank mRNA BC049036.1 - 1 - 2 187 92
BC071429.1 - 1 - 2 164 92

View Full Product Details