Assay Design



Gene Transcripts

Gene Symbol
gcsha
Entrez Gene ID
541329
Gene Name
glycine cleavage system protein H (aminomethyl carrier), a
Gene Aliases
wu:fb14d10, wu:fw84b08, zgc:112535
Chromosome Location
Chr.18: 29489 - 37206 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013457.1 NP_001013475.1 2 - 3 270 90
GenBank mRNA BC091548.1 - 2 - 3 270 90
BC164865.1 - 2 - 3 268 90

View Full Product Details