Assay Design



Gene Transcripts

Gene Symbol
xrcc3
Entrez Gene ID
541414
Gene Name
X-ray repair complementing defective repair in Chinese hamster cells 3
Gene Aliases
im:7142103, si:dkey-11b8.1, zgc:101608
Chromosome Location
Chr.20: 20896690 - 20901329 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013541.1 NP_001013559.1 1 - 2 155 112
GenBank mRNA BC090820.1 - 1 - 2 155 112
BC164394.1 - 1 - 2 90 112

View Full Product Details