Assay Design



Gene Transcripts

Gene Symbol
ahr1b
Entrez Gene ID
554265
Gene Name
aryl hydrocarbon receptor 1b
Gene Aliases
ahr1, si:dkey-159d6.1
Chromosome Location
Chr.22: 12979213 - 13042992 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024816.2 NP_001019987.1 5 - 6 1029 73
GenBank mRNA BC163508.1 - 5 - 6 898 73
DQ016494.1 - 5 - 6 1029 73

View Full Product Details