Assay Design



Gene Transcripts

Gene Symbol
cnih1
Entrez Gene ID
406282
Gene Name
cornichon family AMPA receptor auxiliary protein 1
Gene Aliases
cnih, wu:fc21b06, wu:fi12c05, zgc:112228, zgc:56103
Chromosome Location
Chr.13: 36595631 - 36606932 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033106.1 NP_001028278.1 2 - 3 204 103
GenBank mRNA BC045913.1 - - 199 103
BC100049.1 - 2 - 3 204 103

View Full Product Details