Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040037.1 | NP_001035126.1 | 3 - 4 | 432 | 136 |
XM_017357365.2 | XP_017212854.1 | 3 - 4 | 454 | 136 | |
GenBank mRNA | AB239771.1 | - | 3 - 4 | 432 | 136 |
BC163535.1 | - | 3 - 4 | 420 | 136 | |
DQ424857.1 | - | 3 - 4 | 396 | 136 |