Assay Design



Gene Transcripts

Gene Symbol
sppl2
Entrez Gene ID
325540
Gene Name
signal peptide peptidase-like 2
Gene Aliases
SPPL2B, wu:fc16e01, wu:fc85d12, zgc:136525
Chromosome Location
Chr.2: 37480654 - 37503010 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045244.2 NP_001038709.2 4 - 5 515 112
XM_021478906.1 XP_021334581.1 4 - 5 510 112
XM_021478911.1 XP_021334586.1 4 - 5 510 112
GenBank mRNA AY926457.1 - 4 - 5 499 112
BC044512.1 - 4 - 5 483 112
BC116574.1 - 4 - 5 500 112

View Full Product Details