Assay Design



Gene Transcripts

Gene Symbol
slc8a4a
Entrez Gene ID
568512
Gene Name
solute carrier family 8 member 4a
Gene Aliases
-
Chromosome Location
Chr.21: 27047333 - 27186334 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089419.1 NP_001082888.1 2 - 3 1831 78
XM_005161357.4 XP_005161414.1 4 - 5 2850 78
XM_009295486.3 XP_009293761.1 3 - 4 2758 78
XM_021468890.1 XP_021324565.1 4 - 5 2820 78
GenBank mRNA BC163335.1 - 2 - 3 1856 78
EF470289.1 - 2 - 3 1831 78
EU559290.1 - 2 - 3 1831 78

View Full Product Details