Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110531.1 | NP_001104001.1 | 3 - 4 | 504 | 89 |
XM_005168656.4 | XP_005168713.1 | 4 - 5 | 1103 | 89 | |
XM_009302032.3 | XP_009300307.1 | 4 - 5 | 1103 | 89 | |
GenBank mRNA | EU047918.1 | - | 3 - 4 | 504 | 89 |
HQ222883.1 | - | 3 - 4 | 504 | 89 | |
HQ222884.1 | - | 2 - 3 | 379 | 89 | |
HQ222886.1 | - | 3 - 4 | 438 | 89 | |
HQ222887.1 | - | 3 - 4 | 587 | 89 |