Assay Design



Gene Transcripts

Gene Symbol
tbx4
Entrez Gene ID
30070
Gene Name
T-box transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.15: 99463 - 123113 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130914.2 NP_570989.2 7 - 8 1031 68
GenBank mRNA AF179406.1 - 7 - 8 1031 68
BC162554.1 - 7 - 8 981 68

View Full Product Details