Assay Design



Gene Transcripts

Gene Symbol
wnt8b
Entrez Gene ID
30144
Gene Name
wingless-type MMTV integration site family, member 8b
Gene Aliases
wnt-8b
Chromosome Location
Chr.13: 29993593 - 30006260 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130959.2 NP_571034.1 3 - 4 271 101
GenBank mRNA KM655728.1 - 3 - 4 242 101
U10870.1 - 3 - 4 275 101

View Full Product Details