Assay Design



Gene Transcripts

Gene Symbol
wnt8b
Entrez Gene ID
30144
Gene Name
wingless-type MMTV integration site family, member 8b
Gene Aliases
wnt-8b
Chromosome Location
Chr.13: 29993593 - 30006260 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130959.2 NP_571034.1 5 - 6 543 62
GenBank mRNA KM655728.1 - 5 - 6 514 62
U10870.1 - 5 - 6 547 62

View Full Product Details