Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309399.1 | NP_001296328.1 | 5 - 6 | 1241 | 109 |
NM_001309400.1 | NP_001296329.1 | 5 - 6 | 1241 | 109 | |
NM_152962.3 | NP_694494.2 | 5 - 6 | 1241 | 109 | |
XM_009304303.3 | XP_009302578.1 | 5 - 6 | 1283 | 109 | |
XM_009304304.3 | XP_009302579.1 | 5 - 6 | 1284 | 109 | |
GenBank mRNA | AF389400.2 | - | 4 - 5 | 623 | 109 |
AY197498.1 | - | 4 - 5 | 623 | 109 | |
AY197499.1 | - | 4 - 5 | 623 | 109 | |
AY197500.1 | - | 4 - 5 | 623 | 109 | |
AY197501.1 | - | 4 - 5 | 563 | 109 | |
BC162342.1 | - | 4 - 5 | 623 | 109 | |
CU458752.1 | - | 4 - 5 | 576 | 109 |