Assay Design



Gene Transcripts

Gene Symbol
anxa1b
Entrez Gene ID
353358
Gene Name
annexin A1b
Gene Aliases
anx1b, wu:fa16b09, zgc:109778
Chromosome Location
Chr.5: 65022287 - 65037369 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181759.1 NP_861424.1 6 - 7 575 98
GenBank mRNA AY178794.1 - 6 - 7 575 98
BC092685.1 - 7 - 8 595 98
BC164147.1 - 6 - 7 578 98

View Full Product Details