Assay Design



Gene Transcripts

Gene Symbol
sult1st1
Entrez Gene ID
323424
Gene Name
sulfotransferase family 1, cytosolic sulfotransferase 1
Gene Aliases
fb98b02, fk46b09, wu:fb98b02, wu:fk46b09
Chromosome Location
Chr.8: 46499097 - 46508857 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182941.1 NP_891986.1 3 - 4 384 73
GenBank mRNA AY181064.1 - 3 - 4 384 73
BC056729.1 - 3 - 4 298 73

View Full Product Details