Assay Design



Gene Transcripts

Gene Symbol
timm17a
Entrez Gene ID
386636
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
fb82c12, wu:fb82c12
Chromosome Location
Chr.11: 81541 - 87580 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198367.1 NP_938181.1 2 - 3 131 88
GenBank mRNA BC056525.1 - 2 - 3 131 88
BC065901.1 - 2 - 3 142 88

View Full Product Details