Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199641.1 | NP_955935.1 | 9 - 10 | 1000 | 131 |
XM_005161943.4 | XP_005162000.1 | 10 - 11 | 1109 | 131 | |
XM_005161944.4 | XP_005162001.1 | 10 - 11 | 1571 | 131 | |
XM_005161945.4 | XP_005162002.1 | 10 - 11 | 1414 | 131 | |
XM_005161946.4 | XP_005162003.1 | 10 - 11 | 1030 | 131 | |
XM_009296662.3 | XP_009294937.1 | 10 - 11 | 1009 | 131 | |
XM_009296664.3 | XP_009294939.1 | 10 - 11 | 1012 | 131 | |
XM_009296665.3 | XP_009294940.1 | 10 - 11 | 1109 | 131 | |
GenBank mRNA | BC058064.1 | - | 9 - 10 | 1000 | 131 |
BC091798.1 | - | 10 - 11 | 1497 | 131 |