Assay Design



Gene Transcripts

Gene Symbol
arf1
Entrez Gene ID
114428
Gene Name
ADP-ribosylation factor 1
Gene Aliases
wu:fb33e02, wu:fb78h01
Chromosome Location
Chr.2: 3062737 - 3077027 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201452.2 NP_958860.1 2 - 3 256 124
GenBank mRNA BC044531.1 - 2 - 3 246 124
CD593552.1 - 2 - 3 200 124

View Full Product Details