Assay Design



Gene Transcripts

Gene Symbol
abhd14a
Entrez Gene ID
404598
Gene Name
abhydrolase domain containing 14A
Gene Aliases
zgc:85972
Chromosome Location
Chr.22: 10698595 - 10706873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207066.2 NP_996949.2 2 - 3 401 92
GenBank mRNA BC066291.1 - 2 - 3 352 92
EH433068.1 - 2 - 3 418 92

View Full Product Details