Assay Design



Gene Transcripts

Gene Symbol
dpf2l
Entrez Gene ID
326933
Gene Name
D4, zinc and double PHD fingers family 2, like
Gene Aliases
Dpf2, REQ, wu:fe50b03, zgc:110767
Chromosome Location
Chr.14: 236273 - 245658 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212696.2 NP_997861.2 8 - 9 982 85
GenBank mRNA AY391455.1 - 8 - 9 935 85
BC092130.1 - 8 - 9 982 85
BC165789.1 - 8 - 9 976 85

View Full Product Details