Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214718.1 | NP_999883.1 | 12 - 13 | 1653 | 100 |
XM_005166014.4 | XP_005166071.1 | 14 - 15 | 1864 | 100 | |
XM_021476791.1 | XP_021332466.1 | 13 - 14 | 1703 | 100 | |
GenBank mRNA | AY576997.1 | - | 12 - 13 | 1653 | 100 |
BC066675.1 | - | 13 - 14 | 1819 | 100 |